Disposable samplers, TargetDispo
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5393-4460 5393-4461 5393-4470 5393-4471
300-0249EA 4050 DKK
300-0249 300-0250 300-0251 300-0252
Disposable samplers, TargetDispo
Prøvetagere Prøvetagere til engangsbrug
This disposable TargetDispo sampler is to take a target point sample at a position in the sample material that has previously been precisely determined. Sampler is closed while it pierces into the powder or granulate, when it reaches the sample removal point it is opened and then closed again, in this way a precise target-point sample is obtained.
  • Easy emptying through the opening (Ø 14 mm) in the handle
  • Clean room manufactured
  • Optionally sterilised by gamma rays
  • Cross contamination is completely eliminated