Disposable samplers, DispoPicker
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5393-5540 5393-5541 5393-5550 5393-5551
300-0259EA 2150 DKK
300-0259 300-0260 300-0261 300-0262
Disposable samplers, DispoPicker
Prøvetagere Prøvetagere til engangsbrug
This disposable DispoPicker is used in the sampling of powders and small-grain bulk materials. Because the tip is closed, the sample material cannot trickle back out of the sample lance.
  • Clean room manufactured
  • Optionally sterilised by gamma rays
  • Cross contamination is completely eliminated
Ideally suited for cross-section samples.