Disposable samplers, DispoTube
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5393-6620 5393-6630 5393-6621 5393-6631
300-0267EA 2150 DKK
300-0267 300-0269 300-0268 300-0270
Disposable samplers, DispoTube
Prøvetagere Prøvetagere til engangsbrug
This DispoTube sampler can be used for taking samples of liquids. Its ergonomic handle makes it easier to work and facilitates a good grip even under difficult conditions, for example with gloves.
  • All-layer and target point samples possible
  • Clean room manufactured
  • Optionally sterilised by gamma rays
  • Cross contamination is completely eliminated