Aluminium headspace caps
Leverandør: PerkinElmer
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B0159356 N9303350
PERCB0159356EA 1450 DKK
PERCB0159356 PERCN9303350
Aluminium headspace caps
Låg Låg til kromatografirør
Un-Assembled aluminum caps and septa for all of your analytical and biological applications.
  • Temperature resistance up to 130 °C
  • Very economical and has low resistance to coring