Foam soap, Refresh™ Azure FOAM
Leverandør: Deb-STOKO
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AZU12LTF AZU1L-UK
115-2266EA 468 DKK
115-2266 115-2267
Foam soap, Refresh™ Azure FOAM
Hudpleje Sæber og huden rensemidler
Lightly perfumed gentle foam hand wash for use in washroom areas in offices and public facilities for general removal of dirt and grime.
  • Contains skin conditioners to improve skin hydration and prevent drying
  • Highly acceptable rich foam formula
  • Non irritating to eyes
  • Perfumed
Certificeringer: EU Ecolabel
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904