Hand cleanser, Estesol® Lotion PURE
Leverandør: Deb-STOKO
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PUW1L PUW2LT
115-2328EA 409 DKK
115-2328 115-2329
Hand cleanser, Estesol® Lotion PURE
Hudpleje Sæber og huden rensemidler
Light duty hand cleanser to remove light industrial soilings, synthetic and petroleum-based oils, greases and lubricants. For use in general industrial and food manufacturing and processing environments. Can be used on sensitive skin.
  • Contains glycerine to help prevent skin dryness
  • Mild blend of surfactants for fast and effective hand cleaning
  • Petroleum solvent-free, silicone-free, perfume-free
Certificeringer: EU Ecolabel
Advarsel: *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904