Vacuum wand, C-series
Leverandør: WINDRUSH TECHNOLOGY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
106518 90-CP C002-Y-92-CP C002-Y-90-CP C003-X-95-CP 98-CP C003-Y-99-CP C003-Y-91-CP C002-X-95-CP C003-Y-92-CP C002-Y-98-CP C003-X-96-CP C001-Y-91-CP C002-X-96-CP 99-CP 91-CP C002-Y C003-Y-90-CP
FLUM106518EA 0 DKK
FLUM106518 FLUM90-CP FLUMC002-Y-92-CP FLUMC002-Y-90-CP FLUMC003-X-95-CP FLUM98-CP FLUMC003-Y-99-CP FLUMC003-Y-91-CP FLUMC002-X-95-CP FLUMC003-Y-92-CP FLUMC002-Y-98-CP FLUMC003-X-96-CP FLUMC001-Y-91-CP FLUMC002-X-96-CP FLUM99-CP FLUM91-CP FLUMC002-Y FLUMC003-Y-90-CP
Vacuum wand, C-series
Vacuum håndteringssystemer
This C series wand bodies are manufactured in ESD safe materials including PEEK tips are the safest means of wafer handling. The tips are optimally sized for each wafer diameter, ensuring that contact is both minimal and secure.
  • Polished contact surface provides excellent adhesion to water
  • Fixed tip and swivel joint body styles are available
  • Unique control valve ensures reliable suction and instantaneous release without particle generation