UHPLC columns, Hypersil GOLD™ Vanquish™ aQ
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
25302-202130-V 25302-152130-V 25302-102130-V
555-7975EA 10514.5 DKK
555-7975 555-7974 555-7920
UHPLC columns, Hypersil GOLD™ Vanquish™ aQ
Kromatografikolonner
These reversed phase columns were developed in conjunction with the Vanquish™ UHPLC system to take advantage of the system's extended pressure capabilities and robustness. The result is a new level of separation, speed and throughput that solve analytical challenges of analysing complex matrices using liquid chromatography and LC-MS.
  • High efficiency enables separation of very complex mixtures
  • High efficiency at high flow rates