UHPLC columns, Hypersil GOLD™ Vanquish™ C18
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
25002-202130-V 25002-052130-V 25002-102130-V 25002-152130-V
554-5324EA 10472 DKK
554-5324 554-5321 554-5322 554-5323
UHPLC columns, Hypersil GOLD™ Vanquish™ C18
Kromatografikolonner
These reversed phase columns were developed in conjunction with the Vanquish™ UHPLC system to take advantage of the system's extended pressure capabilities and robustness. The result is a new level of separation, speed and throughput that solve analytical challenges of analysing complex matrices using liquid chromatography and LC-MS.
  • High efficiency enables separation of very complex mixtures
  • High efficiency at high flow rates