VWR®, Spreaders, L-Shaped, Standard Line
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
612-5496EA 1120 DKK
612-5496 612-5497
VWR®, Spreaders, L-Shaped, Standard Line
Spredere Celle Spredere
These PS, sterile, disposable L-shaped spreaders enable even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium.
  • Smooth spreading surface
  • Spreaders are pre-sterilised, eliminating the need for flaming or autoclaving
  • Available individually packed or bulk packed (5 pieces per bag)