Leather gloves, CT402
Leverandør: DELTA PLUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CT402BL/11 CT402BL/10 CT402BL/8 CT402BL/9 CT402BL/7
DELPCT402BL/11EA 605 DKK
DELPCT402BL/11 DELPCT402BL/10 DELPCT402BL/8 DELPCT402BL/9 DELPCT402BL/7
Leather gloves, CT402
Handsker
These gloves feature a goatskin full grain leather palm and a cotton jersey back.
  • American assembly
  • Knitted cotton jersey provides breathability
  • Goatskin thickness: 0,70 to 0,90 mm
Ideal for use in agriculture, construction and logistics.
Certificeringer: EN 388:2016 (2111X)