Overalls, PP, with hood
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
113-9292EA 930 DKK
113-9292 113-9294 113-9296 113-9298 113-9293 113-9289 113-9290 113-9291 113-9301
Overalls, PP, with hood
Engangsbeklædning
Overalls made from non woven PP. Ideal for non critical applications.
  • With hood
  • Elasticated waist, wrist and ankles
  • Zip fastening
  • Latex-free
Weight: 40 g/m²
Colour: White/blue/red