Safety ankle boots, lace-up, Step One, Premiere
Leverandør: U-Power
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SO10104/35 SO10104/36 SO10104/37 SO10104/38 SO10104/39 SO10104/40 SO10104/41 SO10104/42 SO10104/43 SO10104/44 SO10104/45 SO10104/46 SO10104/47 SO10104/48
114-3189EA 639 DKK
114-3189 114-3190 114-3191 114-3192 114-3193 114-3194 114-3195 114-3196 114-3197 114-3198 114-3199 114-3200 114-3201 114-3202
Safety ankle boots, lace-up, Step One, Premiere
Sko og støvler
Safety ankle boots made from water resistant leather with textile inserts and a slip-resistant dual-density PU sole.
  • Comfortable, anatomically shaped Ergo dry insole features a dual PE/EVA support system with textile surface; anti-static and anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Air toe protective aluminium toecap with breathable membrane (200 J)
  • Puncture-resistant steel midsole
Colour: Black
Colour: Black
Certificeringer: EN 20345:2011; S3, SRC