Safety shoes, lace-up, The Roar, Forest
Leverandør: U-Power
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SN20044/35 SN20044/36 SN20044/37 SN20044/38 SN20044/39 SN20044/40 SN20044/41 SN20044/42 SN20044/43 SN20044/44 SN20044/45 SN20044/46 SN20044/47 SN20044/48
114-2892EA 780 DKK
114-2892 114-2893 114-2894 114-2895 114-2896 114-2897 114-2898 114-2899 114-2900 114-2912 114-2913 114-2914 114-2926 114-2927
Safety shoes, lace-up, The Roar, Forest
Sko og støvler
Safety shoes made from soft water repellent leather 'antique style' and a slip-resistant dual-density PU sole.
  • Light non-slip Action dry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Natural fit width 11 for additional comfort
  • Air toe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Grey
Colour: Grey
Certificeringer: EN 20345:2011; S3, SRC