Safety shoes, lace-up, SK Grip, Demon Grip
Leverandør: U-Power
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SG20066/38 SG20066/39 SG20066/40 SG20066/41 SG20066/42 SG20066/43 SG20066/44 SG20066/45 SG20066/46 SG20066/47
UPOWSG20066/38EA 582 DKK
UPOWSG20066/38 UPOWSG20066/39 UPOWSG20066/40 UPOWSG20066/41 UPOWSG20066/42 UPOWSG20066/43 UPOWSG20066/44 UPOWSG20066/45 UPOWSG20066/46 UPOWSG20066/47
Safety shoes, lace-up, SK Grip, Demon Grip
Sko og støvler
100% non-metallic safety shoes made soft suede leather with highly breathable mesh inserts and a lightweight, extremely flexible, slip resistant PU/PU U-Grip sole.
  • Anatomically shaped Polysoft insole made from soft PU, with pockets in the foot support area
  • PU/PU walking sole with 90° angle blocks and mulitlevel stabalising channels to for extremely high anti-slip features; TPU anti twist insert for additional support
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Airtoe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Colour: Black
Certificeringer: EN 20345:2011; S1P, SRC