Safety boots, lace-up, Rock & Roll, Klever UK
Leverandør: U-Power
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RR70334/35 RR70334/36 RR70334/37 RR70334/38 RR70334/39 RR70334/40 RR70334/41 RR70334/42 RR70334/43 RR70334/44 RR70334/45 RR70334/46 RR70334/47 RR70334/48
UPOWRR70334/35EA 673 DKK
UPOWRR70334/35 UPOWRR70334/36 UPOWRR70334/37 UPOWRR70334/38 UPOWRR70334/39 UPOWRR70334/40 UPOWRR70334/41 UPOWRR70334/42 UPOWRR70334/43 UPOWRR70334/44 UPOWRR70334/45 UPOWRR70334/46 UPOWRR70334/47 UPOWRR70334/48
Safety boots, lace-up, Rock & Roll, Klever UK
Sko og støvler
100% metal-free safety boots made from full-grain pull-up water repellent leather with PU scuff cap and a slip-resistant dual-density PU sole.
  • Comfortable, anatomically formed insole ergo dry features a dual PE/EVA support system with textile surface; anti-static and anti-fungal treated
  • Warm fur lining
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Air toe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Colour: Black
Certificeringer: EN 20345:2011; S3, CI, SRC