Safety shoes, lace-up, Rock & Roll, Sun
Leverandør: U-Power
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RR20056/35 RR20056/36 RR20056/37 RR20056/38 RR20056/39 RR20056/40 RR20056/41 RR20056/42 RR20056/43 RR20056/44 RR20056/45 RR20056/46 RR20056/47 RR20056/48
UPOWRR20056/35EA 551 DKK
UPOWRR20056/35 UPOWRR20056/36 UPOWRR20056/37 UPOWRR20056/38 UPOWRR20056/39 UPOWRR20056/40 UPOWRR20056/41 UPOWRR20056/42 UPOWRR20056/43 UPOWRR20056/44 UPOWRR20056/45 UPOWRR20056/46 UPOWRR20056/47 UPOWRR20056/48
Safety shoes, lace-up, Rock & Roll, Sun
Sko og støvler
100% metal-free safety shoes made from soft suede leather and a slip-resistant dual-density PU sole.
  • Comfortable, anatomically formed insole ergo dry features a dual PE/EVA support system with textile surface; anti-static and anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Air toe Composite perforated safety toe cap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Colour: Black
Certificeringer: EN 20345:2011; S1P, SRC