Safety ankle boots, lace-up, concept plus, Gravel
Leverandør: U-Power
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
UA10584/38 UA10584/39 UA10584/40 UA10584/41 UA10584/42 UA10584/43 UA10584/44 UA10584/45 UA10584/46 UA10584/47
UPOWUA10584/38EA 779 DKK
UPOWUA10584/38 UPOWUA10584/39 UPOWUA10584/40 UPOWUA10584/41 UPOWUA10584/42 UPOWUA10584/43 UPOWUA10584/44 UPOWUA10584/45 UPOWUA10584/46 UPOWUA10584/47
Safety ankle boots, lace-up, concept plus, Gravel
Sko og støvler
Safety ankle boots made from smooth water repellent leather and a heat resistant to 300 °C, antislip nitrile rubber sole.
  • Light non-slip action dry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Air toe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex'covers 100% of the sole of the foot
Colour: Black
Colour: Black
Certificeringer: EN 20345:2011; S3, HI, CI, HRO, RS