NMR Capillary tubes, SP Wilmad-LabGlass
Leverandør: SP Industries
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
WG-1364-2-203M WG-1364-1.7 WG-1364-1 WG-1364-1.7-5 WG-1364-2 WG-1364-2.5A WG-1365-1
WILMWG-1364-2-203MEA 223 DKK
WILMWG-1364-2-203M WILMWG-1364-1.7 WILMWG-1364-1 WILMWG-1364-1.7-5 WILMWG-1364-2 WILMWG-1364-2.5A WILMWG-1365-1
NMR Capillary tubes, SP Wilmad-LabGlass
Rør NMR rør
Pyrex® glass, Class A, ASTM type 1.
  • Tested at 400 MHz field
  • Ideal for small volume NMR measurement
  • With either both ends open or sealed at one end