Cover briller, super f OTG
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9169.585 9169.586 9169.615
111-1910EA 276 DKK
111-1910 111-1873 UVEX9169.615
Cover briller, super f OTG
Briller
Super f OTG safety glasses combine all the principles of contemporary safety spectacles with the advantages of state of-the-art lens technology.
  • Ultra-lightweight
  • Slim side arms with soft ends for a pressure-free fit
  • Compatible with earmuffs and safety helmets
  • Fit over most prescription glasses
Ideal for any wearer situation and also can be used as cover spectacles for wearers of prescription glasses.
Certificeringer: EN 166, EN 170