VWR®, Dispensers for Sealing Films
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
631-1137EA 727 DKK
631-1137 631-1138 631-1139
VWR®, Dispensers for Sealing Films
Forseglingsfilm Dispenser
ABS dispenser designed to store, dispense and cut Parafilm™, tape, labels and Tough-Tags™.
  • Design incorporates a safety razor
  • Slides and cuts with no danger to fingers
  • Holds one or two 50 mm (2") rolls, or one 100 mm (4") roll