Safety shoes, lace-up, Concept Plus, Corner
Leverandør: U-Power
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
UC20014/38 UC20014/39 UC20014/40 UC20014/41 UC20014/42 UC20014/43 UC20014/44 UC20014/45 UC20014/46 UC20014/47 UC20014/48
114-3239EA 716 DKK
114-3239 114-3240 114-3241 114-3242 114-3243 114-3244 114-3245 114-3246 114-3247 114-3248 114-3249
Safety shoes, lace-up, Concept Plus, Corner
Sko og støvler
100% metal-free safety shoes made from water repellent full grained leather with abrasive resistant bay-cast protection and a hard wearing, slip resistant PU sole.
  • Light non-slip Actiondry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Airtoe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Certificeringer: EN 20345:2011; S3, SRC