Safety ankle boots, lace-up, Concept Plus, Cross
Leverandør: U-Power
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
UC10014/38 UC10014/39 UC10014/40 UC10014/41 UC10014/42 UC10014/43 UC10014/44 UC10014/45 UC10014/46 UC10014/47 UC10014/48
114-3228EA 794 DKK
114-3228 114-3229 114-3230 114-3231 114-3232 114-3233 114-3234 114-3235 114-3236 114-3237 114-3238
Safety ankle boots, lace-up, Concept Plus, Cross
Sko og støvler
100% metal-free safety ankle boots made from water repellent full grained leather with abrasive resistant bay-cast protection and a hard wearing, slip resistant PU sole.
  • Light non-slip Actiondry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Airtoe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Certificeringer: EN 20345:2011; S3, SRC