Safety ankle boots, lace-up, Concept M, Trail
Leverandør: U-Power
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
UM10034/38 UM10034/39 UM10034/40 UM10034/41 UM10034/42 UM10034/43 UM10034/44 UM10034/45 UM10034/46 UM10034/47
UPOWUM10034/38EA 975 DKK
UPOWUM10034/38 UPOWUM10034/39 UPOWUM10034/40 UPOWUM10034/41 UPOWUM10034/42 UPOWUM10034/43 UPOWUM10034/44 UPOWUM10034/45 UPOWUM10034/46 UPOWUM10034/47
Safety ankle boots, lace-up, Concept M, Trail
Sko og støvler
Anti-static safety ankle boots made from water repellent full grain leather and a heat resistant to 300 °C, antislip nitrile rubber sole.
  • Light non-slip Actiondry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Airtoe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole “Save & Flex”covers 100% of the sole of the foot
Colour: Brown
Certificeringer: EN 20345:2011; S3, SRC, HI, CI, HRO