Cut resistant gloves, uvex unidur 6659 foam
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6093807 6093808 6093809 6093810 6093811
112-4444EA 631 DKK
112-4444 112-4445 112-4446 112-4447 112-4448
Cut resistant gloves, uvex unidur 6659 foam
Handsker
The HPPE and glass fibres of the uvex unidur 6659 foam ensure very high cut protection. These mottled grey safety gloves are particularly dirt-resistant and the NBR foam coating protects against moisture.
  • Excellent cut protection (Cut level 5)
  • High dexterity and flexibility
  • High abrasion resistance
Ideal for use in the automotive and metal work industry, as well as for packaging applications. Suitable for dry and slightly damp areas.
Certificeringer: EN 388:2016 (4×43C)