Safety boots, lace-up, U-Gel, Ice
Leverandør: U-Power
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
UR70134/38 UR70134/39 UR70134/41 UR70134/42 UR70134/43 UR70134/47
UPOWUR70134/38EA 0 DKK
UPOWUR70134/38 UPOWUR70134/39 UPOWUR70134/41 UPOWUR70134/42 UPOWUR70134/43 UPOWUR70134/47
Safety boots, lace-up, U-Gel, Ice
Sko og støvler
Safety ankle boots made from smooth leather with breathable mesh inserts and a shock absorbing TPU sole.
  • Light non-slip Actiondry insole made from EVA/textile keeps feet dry; anti-fungal treated
  • Thinsulate for thermal insulation
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Airtoe protective aluminium toecap with breathable membrane (200 J)
  • Light, flexible and non-metallic puncture resistant midsole 'Save & Flex' covers 100% of the sole of the foot
Colour: Black
Certificeringer: EN 20345:2011; S3, SRC