uvex unidur 6649 Cut Resistant Gloves
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6051608 6051610 6051611 6051609 6051607
112-4500EA 671 DKK
112-4500 112-4502 112-4503 112-4501 112-4499
uvex unidur 6649 Cut Resistant Gloves
Handsker
These mottled grey HPPE/PA/elastane gloves with PU coating are ideal for both clean and dirty environments. The HPPE fibre ensures good cut protection (level 3).
  • Good cut resistance
  • High abrasion resistance, high flexibility
  • Highest level of durability – ideal for challenging environments
  • Outstanding dexterity
Ideal for use in the automotive and metal industry as well as for packaging. Suitable for dry and slightly moist areas.
Certificeringer: EN 388:2016 (4342B).