uvex unilite 6605 Industrial Gloves
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6057307 6057308 6057309 6057310 6057311
112-4439EA 220 DKK
112-4439 112-4440 112-4441 112-4442 112-4443
uvex unilite 6605 Industrial Gloves
Handsker
Lightweight knitted gloves with breathable nitrile foam coating on palm and fingertips.
  • Breathable coating
  • Good mechanical abrasion resistance
  • Good secure grip
  • Very good dexterity right to the fingertips
Ideal for inspection, precision assembly work, small parts assembly and sorting. Suitable for use in dry and slightly damp areas.
Certificeringer: EN 388 (4122).