Safety ankle boots, lace-up, Style & Job, Krotal
Leverandør: U-Power
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BC70463/35 BC70463/36 BC70463/37 BC70463/38 BC70463/39 BC70463/40 BC70463/41 BC70463/42 BC70463/43 BC70463/44 BC70463/45 BC70463/46 BC70463/47 BC70463/48
UPOWBC70463/35EA 403 DKK
UPOWBC70463/35 UPOWBC70463/36 UPOWBC70463/37 UPOWBC70463/38 UPOWBC70463/39 UPOWBC70463/40 UPOWBC70463/41 UPOWBC70463/42 UPOWBC70463/43 UPOWBC70463/44 UPOWBC70463/45 UPOWBC70463/46 UPOWBC70463/47 UPOWBC70463/48
Safety ankle boots, lace-up, Style & Job, Krotal
Sko og støvler
Anti-static safety ankle boots made from water resistant leather and a slip-resistant and hard wearing PU sole.
  • Removable anatomical FlatFit insole
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Colour: Black
Certificeringer: EN 20345:2011; S3, SRC, CI, UK