Pasteur pipettes, disposable, MBL®
Leverandør: SciLabware
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SPP002 SPP004
BIBBSPP002EA 563 DKK
BIBBSPP002 BIBBSPP004
Pasteur pipettes, disposable, MBL®
Pipetter Pasteur-/Overførselspipetter
Soda lime glass, unwadded.
  • Single use Pasteur pipetts for the transfer of small volumes of liquid
  • Pasteur pipettes have a tip internal diameter of 1,1 mm
  • Can be combined with either clear silicone or red latex natural rubber teats (supplied separately)
Certificeringer: Conform to USP Type III requirements.