Safety ankle boots, buckle fastening, Style & Job, Crocodile
Leverandør: U-Power
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BC10423/35 BC10423/36 BC10423/37 BC10423/38 BC10423/39 BC10423/40 BC10423/41 BC10423/42 BC10423/43 BC10423/44 BC10423/45 BC10423/46 BC10423/47 BC10423/48
UPOWBC10423/35EA 456 DKK
UPOWBC10423/35 UPOWBC10423/36 UPOWBC10423/37 UPOWBC10423/38 UPOWBC10423/39 UPOWBC10423/40 UPOWBC10423/41 UPOWBC10423/42 UPOWBC10423/43 UPOWBC10423/44 UPOWBC10423/45 UPOWBC10423/46 UPOWBC10423/47 UPOWBC10423/48
Safety ankle boots, buckle fastening, Style & Job, Crocodile
Sko og støvler
Anti-static safety ankle boots made from water resistant leather and a slip-resistant and hard wearing PU sole.
  • Removable anatomical FlatFit insole
  • Wingtex tunnel lining with honeycomb structure ensures breathability and excellent absorbance features
  • Stress-out tongue system allows easy instep and provides added comfort when walking
  • Natural fit width 11 for additional comfort
  • Protective steel toecap (200 J)
  • Puncture resistant steel midsole
Colour: Black
Certificeringer: EN 20345:2011; S3, SRC