Precision Balances, Scout® STX series
Leverandør: OHAUS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30253058 30253059 30253060 30253061 30253062 30253063 30253064 30253065 30253066 30253067 30253068 30253069
611-3493EA 7550 DKK
611-3493 611-3494 611-3495 611-3496 611-3497 611-3498 611-3499 611-3500 611-3501 611-3502 611-3503 611-3504
Precision Balances, Scout® STX series
Vægte
Ideal for laboratory and industrial applications, the durable Scout® STX series features an easy to navigate colour touchscreen display and a slim design which allows stackable storage with optional storage cover.
  • Portable if used with 4× AA batteries
  • Large, resilient colour touchscreen
  • Milligram balances STX123 and STX223 have an ingenious draught shield with an easy to remove top cover piece
  • Rapid and efficient navigation with the informative icon driven menu structure
  • Fast weighing speed and high resolution deliver repeatable and reliable results
  • Overload protection (10× capacity) and transportation lock
  • Multiple weighing modes: Weighing, parts counting, % weighing, check weighing, dynamic weighing, totalisation , density determination, display hold and mole weighing
The Scout offers multiple built-in application modes, making even the most complex weighing tasks easy to perform. With nine language options, users can even operate the scale in their own language. A real-time clock function keeps accurate time even during power loss. Additionally, GLP/GMP tracking capabilities assist in the monitoring and reporting of data.
Leveringsinformation: Supplied with AC adaptor, and calibration weights are included with models up to 620 g capacity.