Isolator gloves, AlphaTec® CSM
Leverandør: Ansell
Ratings:
Gennemsntilig vurdering: 5.0 (Bedømmelse)
Samlet vurdering
Gennemsntilig vurdering
Var denne bedømmelse brugbar? Ja Nej
Ja: 0 Nej: 0 
Ja, jeg anbefaler
J'aime beaucoup ce produit, je le recommande il est très bien.
85-302/11 85-301/11 85-302/9.5 85-300/11 85-301/9.5 85-300/9.5 85-305/11 85-304/9.5 85-304/11 85-303/9.5 85-303/11 85-305/9.5
112-4972EA 5960 DKK
112-4972 112-4970 112-4973 112-4968 112-4971 112-4969 112-4978 112-4977 112-4976 112-4975 112-4974 112-4979
Isolator gloves, AlphaTec® CSM
Handsker
These ambidextrous isolator box gloves are made ​​of chlorosulfonated polyethylene (CSM) without an inner lining. They are designed for high resistance against concentrated acids and bases.
  • Soft, flexible material designed for ease of use
  • Resists ageing from exposure to oxygen, UV rays and ozone
  • Withstands temperatures up to 120 °C
  • White colouring for easy detection of contamination
  • Heavy-weight version with increased thickness (0,6 mm) for added durability
Ideal for pharmaceutical applications.
Certificeringer: EN ISO 374-1 Type B (AMP), EN 374-5 VIRUS, EN 388 (3101A); Cat. III.