VWR® Talon®, Clamps, Three Pronged
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
241-0404EA 883 DKK
241-0404 241-0405 241-0406
VWR® Talon®, Clamps, Three Pronged
Klemmer Universalklemmer
Stainless steel or nickel-plated zinc. These clamps securely hold every type of glassware and apparatus.
  • Extension arms allow placement of apparatus at various distances from lab frames without compromising the integrity of the experiment
  • Stainless steel clamps: Exceptional chemical resistance, clamps are electropolished and autoclavable
  • Nickel-plated zinc clamps: High tensile strength
  • Supplied with non-slip vinyl sleeves, and fiberglass covers for temperatures above 100 °C
Long, seamless tubing attaches clamp head securely and offers easy positioning in the deepest fume hoods. Clamps are constructed with round extension arms that allow the clamps to be rotated 360°.
Bestillingsoplysninger: An additional holder must be purchased in order to attach clamps to frames or other apparatus.