GPC calibration mix
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
32019 32023
REST32019EA 314 DKK
REST32019 REST32023
GPC calibration mix
Standarder Økologiske standarder
These qualitative mixtures are useful for determining GPC dump/collect times. The compounds are dissolved in methylene chloride.
This mix includes bis (2-ethylhexyl) phthalate (10 mg/ml), perylene (0,2 mg/ml), corn oil (250 mg/ml), sulphur (0,8 mg/ml), methoxychlor (2,0 mg/ml), in methylene chloride.