Safety disposal cans, quick-disconnect
Leverandør: JUSTRITE MANUFACTURING
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PP12755 TF12752 TF12755 PP12752 BY12752 BY12755 12160
119-0439EA 3610 DKK
119-0439 119-0440 119-0441 119-0438 119-0436 119-0437 119-0435
Safety disposal cans, quick-disconnect
Sikkerhedsdunke
Quick-disconnect safety disposal cans are the safe, compliant way to collect HPLC waste. Standardized configurations with a choice of stainless steel or polypropylene fittings or a combination of the two.
PE.
  • Hands-free solvent collection
  • Secure, easy connection and disconnection
  • Durable and safe with a maximum usage of manifolds enable upto 12 waste lines to be fed into a single intake fitting