Coliform water testing vials, wide mouth, Corning®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1700-100 1705-100
CORN1700-100EA 694 DKK
CORN1700-100 CORN1705-100
Coliform water testing vials, wide mouth, Corning®
Hætteglas
These graduated, sterile, cylindrical vials made from PP are used in testing for the presence of coliform bacteria, microbiological contaminants in drinking water.
  • One-piece container with attached lid to reduce chance of contamination
  • Locking arrow ensures sterility
  • Leak tight when sealed properly
  • Tie-down to protect from accidental opening
  • With or without sodium thiosulfate tablet
Meets EPA requirements.
Leveringsinformation: Sample label and instructions for use are supplied.
Advarsel: Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904.