Crimp/snap top autosampler vials
Leverandør: THERMO NATIONAL SCIENTIFIC
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C4011-14 C4011-13
NSCAC4011-14EA 0 DKK
NSCAC4011-14 NSCAC4011-13
Crimp/snap top autosampler vials
Hætteglas
Chemically inert PP vials, manufactured from high quality, GC tested PP material.
  • Alternative to glass for some applications
  • Chemically inert and suitable for most chromatography applications
  • Can be used when levels of inorganic ions have to be kept to an absolute minimum