Ribbons for Signature® slide printer
Leverandør: Primera Technology Europe
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
058105 058106
720-2879EA 0 DKK
720-2879 720-2880
Ribbons for Signature® slide printer
Printer bånd
Ribbons come in Standard, TC1 or TC2 quality.
  • Standard prints on many smooth surfaces and is bath resistant against chemicals but is not wipe resistant
  • TC1 prints on selected slides and is wipe resistant against chemicals as well temperature resistant over 95 degree Celsius up to 110 degrees
  • TC2 has a wide compatibility to various slide surfaces and is touch resistant against chemicals
VWR® recommend to test the proper combination before buying large slide quantities.