Gas-tight suits, Tychem®10000 TK, with attached boots
Leverandør: DuPont
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TYKGEVJDYL00/L TYKGEVJDYL00/M TYKGEVJDYL00/S TYKGEVJDYL00XL TYKGEVJDYL0XXL
DUPOTYKGEVJDYL00/LEA 32800 DKK
DUPOTYKGEVJDYL00/L DUPOTYKGEVJDYL00/M DUPOTYKGEVJDYL00/S DUPOTYKGEVJDYL00XL DUPOTYKGEVJDYL0XXL
Gas-tight suits, Tychem®10000 TK, with attached boots
Engangsbeklædning
This encapsulated gas-tight garment with detachable Hazmax boots (sizes 6 to 14 (39 to 48)) exhibits excellent chemical barrier properties. It is made from an extremely durable fabric that is puncture- and tear-resistant, yet also supple and lightweight (<4,6 kg per garment).
  • Wide, anti-mist visor for undistorted, panoramic visibility
  • Bat-wing design to allow the wearer to withdraw an arm to attend to breathing apparatus
  • Internal, adjustable waist belt system for support and improved fit
  • Attached double gloves, five-finger, with locking cuff mechanism for glove replacement
  • Double-taped seams provide strong chemical resistance against heavy liquid splashes
  • Heavy-duty gas-tight zipper that is extra-long to facilitate garment opening and aid donning and doffing
  • Overlapping zipper flaps with hook and loop closures
Tychem® 10000 TK. fabric shows at least 30 minutes of barrier protection to more than 300 challenge chemicals. The garment is specifically developed for protection against toxic, corrosive gases, liquids and solid chemicals and is suited for industrial, HazMat and domestic preparedness applications.
Each suit has a unique serial number and is fully tested at time of manufacture, including positive air pressure integrity testing.
Colour: Lime Green
Shelf life: 5 years (when stored correctly)
Please note: Sizes are MTO: Made to order terms & conditions apply.
Certificeringer: Type 1a-ET, limited use, EN 943-2 (protective clothing against liquid and gaseous chemicals); Cat. III