Buffer tablets according to Weise, Sigma-Aldrich®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.09468.0100 1.11374.0100 1.11373.0100
1.09468.0100EA 868 DKK
1.09468.0100 1.11374.0100 1.11373.0100
Buffer tablets according to Weise, Sigma-Aldrich®
Buffere Mikroskopi buffere
Ideal for preparing buffer solution according to WEISE for staining of blood smears.
Prepared buffer solutions are used in hematology, for the dilution of staining solutions and for the rinsing steps in the staining process. Each tablet is sufficient for one litre of buffer solution, each produced solution can be stable for up to 4 weeks.