Disposable samplers, ViscoDispo
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5393-3320 5393-3330 5393-3331 5393-3321
300-0206EA 2860 DKK
300-0206 300-0208 300-0209 300-0207
Disposable samplers, ViscoDispo
Prøvetagere Prøvetagere til engangsbrug
This ViscoDispo sampler is for highly viscous media up to 100 000 cP. The ergonomically shaped handle and stable drawbar make the sample easier to draw.
  • Clean room manufactured
  • Optionally sterilised by gamma rays
  • Cross contamination is completely eliminated