Disposable samplers, MicroDispo
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5393-2251 5393-2240 5393-2250 5393-2241
300-0242EA 4930 DKK
300-0242 300-0238 300-0241 300-0239
Disposable samplers, MicroDispo
Prøvetagere Prøvetagere til engangsbrug
The disposable MicroDispo samplers can be used for pourable bulk goods up to 0,5 mm particle size.
  • Cleanroom manufactured
  • Sterilised models are gamma irradiated
  • Cross contamination is completely eliminated
Using this sampler, the precise target point samples can be taken of free- flowing powders and small-grain bulk materials.