Disposable samplers, DispoPipette
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5393-5531 5393-5532 5393-5500 5393-5501
300-0257EA 2210 DKK
300-0257 300-0258 300-0253 300-0254
Disposable samplers, DispoPipette
Prøvetagere Prøvetagere til engangsbrug
Disposable sampler for the highest standard in sampling unlike conventional reusable samplers, disposable samplers do not need to be cleaned after use. Simple disposable sampler for taking samples of low viscosity media. Cleaning reusable samplers is a demanding and often complicated task with greasy toxic substances. Sometimes the task is even impossible. Cross contamination is completely eliminated. It has two opening sizes (2,5/5,5 mm) for different viscosities.
  • All-layer and target point samples are possible
  • Clean room manufactured
  • Optionally sterilised by gamma rays