Disposable samplers, PowderDispo
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5393-5521 5393-5522 5393-5510 5393-5511
300-0211EA 3110 DKK
300-0211 300-0210 300-0255 300-0256
Disposable samplers, PowderDispo
Prøvetagere Prøvetagere til engangsbrug
These disposable samplers for the highest standards do not need to be cleaned after use.
  • Cleanroom manufactured
  • Optionally sterilised by gamma rays
  • Cross contamination is completely eliminated
The PowderDispo is a disposable sampler for cross-section samples of powdery or slightly cohesive substances. A complete all-layer samples can be taken from bags or smaller containers with the lance.