Disposable samplers, LiquiDispo
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5393-1120 5393-1130 5393-1131 5393-1121
300-0202EA 2860 DKK
300-0202 300-0204 300-0205 300-0203
Disposable samplers, LiquiDispo
Prøvetagere Prøvetagere til engangsbrug
Cleaning reusable samplers is a demanding and often complicated task, with greasy, strongly adhesive or toxic substances. An ergonomic handle makes it easier to draw the sample as with a syringe.
  • Ergonomic handle, no seals
  • Cleanroom manufactured
  • Optionally sterilised by gamma rays
  • Cross contamination is completely eliminated
Used to take sample of liquids from highly liquid to slightly viscous.