Inserts for screw neck vials
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
21775 24508 24510 24511 24512 24691
REST21775EA 6010 DKK
REST21775 REST24508 REST24510 REST24511 REST24512 REST24691
Inserts for screw neck vials
Hætteglas
These inserts can be used with 2 ml, 8 mm screw neck vials.