Capillary column nuts for Agilent GCs
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20883 21884 21878 21879
REST20883EA 366 DKK
REST20883 REST21884 REST21878 REST21879
Capillary column nuts for Agilent GCs
Fittings og stik
These nuts are designed for Agilent 5890/6850/6890/7890 GCs. Nuts for use with standard ¹/₁₆" type ferrules are designed to fit a wider variety of ¹/₁₆" ferrules.