Precision thermometers, low temperature
Leverandør: Ludwig Schneider GmbH & CO. KG
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1042402 1052006 1052005 1052004 1052003 1042602 1042504 1042403 1042405 1052009 1052104
SCNL1042402EA 665 DKK
SCNL1042402 SCNL1052006 SCNL1052005 SCNL1052004 SCNL1052003 SCNL1042602 SCNL1042504 SCNL1042403 SCNL1042405 SCNL1052009 SCNL1052104
Precision thermometers, low temperature
Termometre Analoge termometre
These precision, low temperature laboratory thermometers are suitable for official certification.
  • Total immersion
  • Colourless prism or yellow back round capillary
Pentane, alcohol or toluene, enlcosed or stem type.
Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.
Bestillingsoplysninger: Available with works certificate or with DAkkS calibration certificate (optional).