Syringe filters, with Luer lock inlet
Leverandør: Restek
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
23975 23974 26149 23984 23985 28935
REST23975EA 814 DKK
REST23975 REST23974 REST26149 REST23984 REST23985 REST28935
Syringe filters, with Luer lock inlet
Filtre Sprøjtefiltre
Syringe filters with hydrophobic PTFE membranes and PP housing are used for organic solvents, acids, alcohols, bases and aromatics.
  • Luer lock inlet for leak-tight syringe connection
  • Autoclavable at 121 °C for 15 min
  • Colour coded for easy identification